missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL34 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97371B
This item is not returnable.
View return policy
Description
MRPL34 Polyclonal specifically detects MRPL34 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| MRPL34 | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| L34mtMGC24974, MGC2633, mitochondrial ribosomal protein L34, MRP-L34,39S ribosomal protein L34, mitochondrial | |
| Produced in rabbits immunized with E. coli-derived Human MRPL34 fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 64981 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| Biotin | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction