missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL37 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544650
This item is not returnable.
View return policy
Description
Peptide sequence: VRSTRKSEPP PLDRVYEIPG LEPITFAGKM HFVPWLARPI FPPWDRGYKD Sequence homology: Cow: 100%; Dog: 86%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit.
Specifications
| MRPL37 | |
| Polyclonal | |
| Unconjugated | |
| MRPL37 | |
| 2300004O14Rik; 39S ribosomal protein L2, mitochondrial; 39S ribosomal protein L37, mitochondrial; AI132596; HSPC235; L2mt; L37mt; MGC878; Mitochondrial large ribosomal subunit protein mL37; mitochondrial ribosomal protein L37; MRPL2; MRP-L2; MRPL37; MRP-L37; ribosomal protein, mitochondrial, L2; RPML2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51253 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BZE1 | |
| MRPL37 | |
| Synthetic peptide directed towards the N-terminal of human MRPL37 (aa 35-84). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur