missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL38 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 237.00 - € 470.00
Specifications
| Antigen | MRPL38 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618231
|
Novus Biologicals
NBP2-93579-0.02ml |
0.02 mL |
€ 237.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18626570
|
Novus Biologicals
NBP2-93579-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRPL38 Polyclonal antibody specifically detects MRPL38 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| MRPL38 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Endocrinology | |
| PBS (pH 7.3), 50% glycerol | |
| 64978 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| FLJ13996, HSPC262, KIAA1863, L38MT, MGC4810, mitochondrial ribosomal protein L38,39S ribosomal protein L38, mitochondrial, MRP-L3, MRP-L38, RPML3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 35-380 of human MRPL38 (NP_115867.2). MPNSDIDLSNLERLEKYRSFDRYRRRAEQEAQAPHWWRTYREYFGEKTDPKEKIDIGLPPPKVSRTQQLLERKQAIQELRANVEEERAARLRTASVPLDAVRAEWERTCGPYHKQRLAEYYGLYRDLFHGATFVPRVPLHVAYAVGEDDLMPVYCGNEVTPTEAAQAPEVTYEAEEGSLWTLLLTSLDGHLLEPDAEYLHWLLTNIPGNRVAEGQVTCPYLPPFPARGSGIHRLAFLLFKQDQPIDFSEDARPSPCYQLAQRTFRTFDFYKKHQETMTPAGLSFFQCRWDDSVTYIFHQLLDMREPVFEFVRPPPYHPKQKRFPHRQPLRYLDRYRDSHEPTYGIY | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title