missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL38 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145094
This item is not returnable.
View return policy
Description
Immunogen sequence: HRLAFLLFKQ DQPIDFSEDA RPSPCYQLAQ RTFRTFDFYK KHQETMTPAG LSFFQCRWDD SVTYIFHQLL Antigen sequence identity: Mouse 87%; Rat 86%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
Specifications
| MRPL38 | |
| Polyclonal | |
| Unconjugated | |
| MRPL38 | |
| 1110036N21Rik; 39S ribosomal protein L38, mitochondrial; 4733401F03Rik; HSPC262; L38mt; Mitochondrial large ribosomal subunit protein mL38; mitochondrial ribosomal protein L38; MRP-L3; MRPL38; MRP-L38; R75240; ribosomal protein, mitochondrial, L3; rm38; Rpml3 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64978 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96DV4 | |
| MRPL38 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 266-335. Recombinant protein control fragment (Product #RP-109991). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu