missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL39 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | MRPL39 |
|---|---|
| Dilution | Western Blot 1:1000-1:4000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697902
|
Novus Biologicals
NBP2-95123-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689031
|
Novus Biologicals
NBP2-95123-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRPL39 Polyclonal antibody specifically detects MRPL39 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MRPL39 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 54148 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:4000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C21orf92PRED66, FLJ20451,39S ribosomal protein L39, mitochondrial, L39mt39S ribosomal protein L5, mitochondrial, MGC104174, MGC3400, mitochondrial ribosomal protein L39, MRP-L5L5mt, MRPL5MSTP003, PRED22, RPML5MRP-L39 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 39-338 of human MRPL39 (NP_059142.2). ELTEMRNDLFNKEKARQLSLTPRTEKIEVKHVGKTDPGTVFVMNKNISTPYSCAMHLSEWYCRKSILALVDGQPWDMYKPLTKSCEIKFLTFKDCDPGEVNKAYWRSCAMMMGCVIERAFKDEYMVNLVRAPEVPVISGAFCYDVVLDSKLDEWMPTKENLRSFTKDAHALIYKDLPFETLEVEAKVALEIFQHSKYKVDFIEEKASQNPERIVKLHRIGDFIDVSEGPLIPRTSICFQYEVSAVHNLQPTQPSLIRRFQGVSLPVHLRAHFTIWDKLLERSRKMVTEDQSKATEECTST | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title