missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL39 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-19406
This item is not returnable.
View return policy
Description
MRPL39 Polyclonal specifically detects MRPL39 in Human samples. It is validated for Western Blot.
Specifications
| MRPL39 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| C21orf92PRED66, FLJ20451,39S ribosomal protein L39, mitochondrial, L39mt39S ribosomal protein L5, mitochondrial, MGC104174, MGC3400, mitochondrial ribosomal protein L39, MRP-L5L5mt, MRPL5MSTP003, PRED22, RPML5MRP-L39 | |
| Rabbit | |
| 39 kDa | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 54148 | |
| Human | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9NYK5 | |
| MRPL39 | |
| Recombinant protein encompassing a sequence within the center region of human MRPL39. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Bovine (91%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction