missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL42 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97198IR
This item is not returnable.
View return policy
Description
MRPL42 Polyclonal antibody specifically detects MRPL42 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MRPL42 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| 39S ribosomal protein L31, mitochondrial, 39S ribosomal protein L42, mitochondrial, HSPC204,28S ribosomal protein S32, mitochondrial, L31mt, mitochondrial ribosomal protein L42, mitochondrial ribosomal protein S32, MRPL31L42mt, MRP-L31MRP-S32, MRPS32MRP-L42, PTD007, RPML31S32mt | |
| Produced in rabbits immunized with E. coli-derived Human MRPL42 fragment. | |
| 0.1 mL | |
| Stem Cell Markers | |
| 28977 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction