missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL43 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103527
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MRPL43.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. This gene and the gene for a semaphorin class 4 protein (SEMA4G) overlap at map location 10q24.31 and are transcribed in opposite directions. Sequence analysis identified multiple transcript variants encoding at least four different protein isoforms.
Specifications
| MRPL43 | |
| Polyclonal | |
| Unconjugated | |
| MRPL43 | |
| 39S ribosomal protein L43, mitochondrial; 4930442D21Rik; bMRP36a; L43mt; Mitochondrial large ribosomal subunit protein mL43; Mitochondrial ribosomal protein bMRP36a; mitochondrial ribosomal protein L43; MRPL43; MRP-L43; MRPL43 protein; putative; retina-specific 15.7 kDa protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 309440, 84545, 94067 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N983, Q99N89 | |
| MRPL43 | |
| A synthesized peptide derived from human MRPL43(Accession Q8N983), corresponding to amino acid residues A152-C202. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction