Learn More
MRPL44 Monoclonal Antibody (OTI1A1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810592S
This item is not returnable.
View return policy
Description
MRPL44 Monoclonal antibody specifically detects MRPL44 in Human samples. It is validated for Western BlotSpecifications
| MRPL44 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H9J2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 65080 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1A1 | |
| Unconjugated | |
| MRPL44 | |
| 1810030E18Rik; 39S ribosomal protein L44, mitochondrial; 5730593H20Rik; AI463302; COXPD16; l44; L44mt; Mitochondrial large ribosomal subunit protein mL44; mitochondrial ribosomal protein L44; MRPL44; MRP-L44; RM44 | |
| Full length human recombinant protein of human MRPL44 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |