Learn More
MRPL44 Mouse anti-Human, Clone: OTI2A8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810591
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.Specifications
| MRPL44 | |
| Monoclonal | |
| Unconjugated | |
| MRPL44 | |
| COXPD16, L44MT, MRP-L44 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 65080 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI2A8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9H9J2 | |
| MRPL44 | |
| Full length human recombit protein of human MRPL44 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.