missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPL47 Polyclonal Antibody

Product Code. 15765891 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15765891 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15765891 Supplier Invitrogen™ Supplier No. PA558002

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LKERNMLLTL EQEAKRQRLP MPSPERLDKV VDSMDALDKV VQEREDALRL LQTGQERARP GAWRRDIFGR IIWHKFKQWV I Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. This gene is immediately adjacent to the gene for BAF complex 53 kDa subunit protein a (BAF53a), in a tail-to-tail orientation. Two transcript variants encoding different protein isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL47
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MRPL47
Gene Accession No. Q9HD33
Gene Alias 39S ribosomal protein L47, mitochondrial; 4833424P18Rik; CGI-204; ENSMUSG00000051761; Gm9859; L47mt; metanephros-derived tubulogenic factor; Mitochondrial large ribosomal subunit protein uL29m; mitochondrial ribosomal protein L47; Mrpl47; MRP-L47; MTF/L47; nasopharyngeal carcinoma metastasis-related 1; nasopharyngeal carcinoma metastasis-related protein 1; NCM1
Gene Symbols MRPL47
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRPL47. Recombinant protein control fragment (Product #RP-96340).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57129
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.