missing translation for 'onlineSavingsMsg'
Learn More

MRPL48 Monoclonal Antibody (OTI7F4), TrueMAB™, OriGene

Product Code. 17574374
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17574374 30 μL 30µL
17554374 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17574374 Supplier Origene Technologies Supplier No. TA810857S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MRPL48 Monoclonal antibody specifically detects MRPL48 in Human, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen MRPL48
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7F4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MRPL48
Gene Accession No. Q96GC5
Gene Alias 1810030E20Rik; 2610028L11Rik; 39S ribosomal protein L48, mitochondrial; CGI-118; D4Ertd786e; HSPC290; L48mt; MGC13323; mitochondrial 39S ribosomal protein L48-like protein; Mitochondrial large ribosomal subunit protein mL48; mitochondrial ribosomal protein L48; Mrpl48; MRP-L48; Unknown (protein for MGC:133957)
Host Species Mouse
Immunogen Full length human recombinant protein of human MRPL48 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 293149, 51642
Target Species Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.