missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL48 Monoclonal Antibody (OTI8E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810858S
This item is not returnable.
View return policy
Description
MRPL48 Monoclonal antibody specifically detects MRPL48 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| MRPL48 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96GC5 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51642 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8E1 | |
| Unconjugated | |
| MRPL48 | |
| 1810030E20Rik; 2610028L11Rik; 39S ribosomal protein L48, mitochondrial; CGI-118; D4Ertd786e; HSPC290; L48mt; MGC13323; mitochondrial 39S ribosomal protein L48-like protein; Mitochondrial large ribosomal subunit protein mL48; mitochondrial ribosomal protein L48; Mrpl48; MRP-L48; Unknown (protein for MGC:133957) | |
| Full length human recombinant protein of human MRPL48 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction