missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPL51 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16537981
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16537981 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16537981 Supplier Invitrogen™ Supplier No. PA590748

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MAGNLLSGAG RRLWDWVPLA CRSFSLGVPR LIGIRLTLPP PKVVDRWNEK RAMFGVYDNI GILGNFEKHP KELIRGPIWL RGWKGNELQR CIRKRKMVGS RMFADDLHNL NKRIRYLYKH FNRHGKFR; Positive Samples: Mouse kidney, Rat heart, Rat kidney; Cellular Location: Mitochondrion.

Mammalian mitochondrial ribosomes (mitoribosomes) are responsible for protein synthesis within the mitochondrion. The mitoribosomes are composed of a 4:1 ratio of protein to RNA, with the proteins forming two subunits, the 28S subunit and the 39S subunit. Across species, the proteins that make up the mitoribosome subunits vary greatly in sequence, preventing easy recognition by sequence homology. MRP-L51(mitochondrial ribosomal protein L51), also known as CDA09, MRP64, bMRP64 or HSPC241, is a 128 amino acid mitochondrial ribosomal protein. MRP-L51 is a component of the mitochondrial ribosome large subunit (39S) which consists of a 16S rRNA and about 50 distinct proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRPL51
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.28 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MRPL51
Gene Accession No. Q9CPY1
Gene Alias 2610511M02Rik; 39S ribosomal protein L51, mitochondrial; bMRP64; bMRP-64; CDA09; HSPC241; L51mt; Mitochondrial large ribosomal subunit protein mL51; mitochondrial ribosomal protein 64; mitochondrial ribosomal protein bMRP64; mitochondrial ribosomal protein L51; MRP64; MRPL51; MRP-L51; rm51
Gene Symbols MRPL51
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-128 of human MRPL51 (NP_0575812).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 297601, 66493
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.