missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL52 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06249-50ul
This item is not returnable.
View return policy
Description
MRPL52 Polyclonal antibody specifically detects MRPL52 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MRPL52 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| L52mt, mitochondrial ribosomal protein L52, MRP-L52,39S ribosomal protein L52, mitochondrial | |
| Produced in rabbits immunized with E. coli-derived Human MRPL52 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 122704 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction