missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPL55 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555671
This item is not returnable.
View return policy
Description
Immunogen sequence: PVLLVKQDGS TIHIRYREPR RMLAMPIDLD TLSPEERRAR LRKREAQLQS RKEYEQELSD DLHVERYRQF WTRTKK Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPL55 encodes a 39S subunit protein. Multiple transcript variants encoding two different isoforms were identified through sequence analysis.
Specifications
| MRPL55 | |
| Polyclonal | |
| Unconjugated | |
| MRPL55 | |
| 2810038N09Rik; 39S ribosomal protein L55, mitochondrial; AAVG5835; L55mt; L55nt; Mitochondrial large ribosomal subunit protein bL31m; Mitochondrial large ribosomal subunit protein mL55; mitochondrial ribosomal protein L55; Mrpl55; MRP-L55; PRO19675; Unknown (protein for MGC:137378); UNQ5835/PRO19675 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 128308 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z7F7 | |
| MRPL55 | |
| Recombinant protein corresponding to Human MRPL55. Recombinant protein control fragment (Product #RP-94419). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction