missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRPL9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 17401743 Alle ansehen Thermo Scientific Produkte
Change view
Click to view available options
Quantity:
100 μL
Packungsgröße:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity unitSize
17401743 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Catalog No. 17401743 Lieferant Invitrogen™ Lieferanten-Nr. PA5118078

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

MRPL9 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP

Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein. A pseudogene corresponding to this gene is found at 8q21.11.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen MRPL9
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.03% ProClin 300; pH 7
Gene MRPL9
Gene Accession No. Q9BYD2
Gene Alias 39S ribosomal L9; 39S ribosomal protein L9, mitochondrial; 8030480E20Rik; AA409733; C330013D18Rik; L9mt; Mitochondrial large ribosomal subunit protein bL9m; mitochondrial ribosomal protein L9; Mrpl9; MRP-L9
Gene Symbols MRPL9
Host Species Rabbit
Immunogen E. coli-derived Human MRPL9 fragment.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65005
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.