missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS15 Antibody (OTI5F5), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-44708
This item is not returnable.
View return policy
Description
MRPS15 Monoclonal antibody specifically detects MRPS15 in Human samples. It is validated for Western Blot
Specifications
| MRPS15 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized powder ( 1X PBS, pH 7.3,) | |
| Mouse | |
| >95% | |
| RUO | |
| 64960 | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5F5 | |
| Western Blot 1:2000 | |
| FLJ11564,28S ribosomal protein S15, mitochondrial, mitochondrial ribosomal protein S15, MPR-S15, MRP-S15, RPMS15, S15mt | |
| Full length human recombinant protein of human MRPS15(NP_112570) produced in E.coli. | |
| 100 μg | |
| Primary | |
| For reconstitution, we recommend adding 100uL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiment, we strongly recommend performing another round of desalting process. | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction