missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS15 Monoclonal Antibody (OTI6D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808238
This item is not returnable.
View return policy
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. MRPS15 encodes a 28S subunit protein that belongs to the ribosomal protein S15P family. The encoded protein is more than two times the size of its E. coli counterpart, with the 12S rRNA binding sites conserved. Between human and mouse, the encoded protein is the least conserved among small subunit ribosomal proteins. Pseudogenes corresponding to this gene are found on chromosomes 15q and 19q.Specifications
| MRPS15 | |
| Monoclonal | |
| Unconjugated | |
| MRPS15 | |
| DC37, MPR-S15, RPMS15, S15mt | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64960 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI6D2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P82914 | |
| MRPS15 | |
| Full length human recombit protein of human MRPS15 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction