missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS18B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562171
This item is not returnable.
View return policy
Description
Immunogen sequence: ASVLNTVLRR LPMLSLFRGS HRVQVPLQTL CTKAPSEEDS LSSVPISPYK DEPWKYLESE EYQERYGSRP VWADYRRNHK GGVPPQRTRK Highest antigen sequence identity to the following orthologs: Mouse - 69%, Rat - 71%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S18P family. The encoded protein is one of three that has significant sequence similarity to bacterial S18 proteins. The primary sequences of the three human mitochondrial S18 proteins are no more closely related to each other than they are to the prokaryotic S18 proteins. Pseudogenes corresponding to this gene are found on chromosomes 1q and 2q.
Specifications
| MRPS18B | |
| Polyclonal | |
| Unconjugated | |
| MRPS18B | |
| 2400002C15Rik; 28S ribosomal protein S18-2, mitochondrial; 28S ribosomal protein S18b, mitochondrial; C6orf14; HSPC183; HumanS18a; mitochondrial ribosomal protein S18-2; mitochondrial ribosomal protein S18B; Mitochondrial small ribosomal subunit protein bS18b; Mitochondrial small ribosomal subunit protein mS40; MRP-S18-2; MRPS18-2; MRPS18B; MRP-S18-b; mrps18-b; PTD017; S18amt; S18mt-b | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 28973 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y676 | |
| MRPS18B | |
| Recombinant protein corresponding to Human MRPS18B. Recombinant protein control fragment (Product #RP-100645). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction