missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS2 Antibody (OTI4D6), PE/Atto594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72783PEATT594
This item is not returnable.
View return policy
Description
MRPS2 Monoclonal specifically detects MRPS2 in Human samples. It is validated for Flow Cytometry.
Specifications
| MRPS2 | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| MRPS2 | |
| Full length recombinant protein of human MRPS2 (NP_057118) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| OTI4D6 | |
| Flow Cytometry | |
| 28S ribosomal protein S2, mitochondrial, CGI-91, mitochondrial 28S ribosomal protein S2, mitochondrial ribosomal protein S2, MRP-S2, S2mt | |
| Mouse | |
| Affinity Purified | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51116.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction