missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS25 Antibody (38E7), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-42628UV
This item is not returnable.
View return policy
Description
MRPS25 Monoclonal specifically detects MRPS25 in Human samples. It is validated for Western Blot, ELISA.
Specifications
| MRPS25 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MRPS25 | |
| Recombinant human MRPS25 (1-173aa) purified from E. coli | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 64432 | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 380000000 | |
| Western Blot, ELISA | |
| FLJ00023, mitochondrial 28S ribosomal protein S25, mitochondrial ribosomal protein S25, MRP-S2528S ribosomal protein S25, mitochondrial, RPMS25DKFZp313H0817, S25mt | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction