missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112180
This item is not returnable.
View return policy
Description
MRPS26 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), IP
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. This gene lies adjacent to and downstream of the gonadotropin-releasing hormone precursor gene.
Specifications
| MRPS26 | |
| Polyclonal | |
| Unconjugated | |
| MRPS26 | |
| 28S ribosomal protein S13, mitochondrial; 28S ribosomal protein S26, mitochondrial; 5'OT-EST protein; AI648866; C20orf193; dJ534B8.3; GI008; mitochondrial ribosomal protein S26; Mitochondrial small ribosomal subunit protein mS26; MRPS13; MRP-S13; Mrps26; MRP-S26; NY-BR-87; protein 5'OT-EST; RPMS13; rt26; S13mt; S26mt; serologically defined breast cancer antigen NY-BR-87 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 64949 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q9BYN8 | |
| MRPS26 | |
| E. coli-derived Human MRPS26 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction