missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS27 Mouse anti-Human, Clone: 1B1B3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66724-1-IG-150UL
This item is not returnable.
View return policy
Description
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that may be a functional partner of the death associated protein 3 (DAP3).Specifications
| MRPS27 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q92552 | |
| MRPS27 | |
| MRPS27 Fusion Protein Ag11047 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1B1B3 | |
| Unconjugated | |
| MRPS27 | |
| FLJ21764, FLJ23348, KIAA0264, MRP S27, MRPS27, S27mt | |
| Mouse | |
| Protein A | |
| RUO | |
| 23107 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction