Learn More
MRPS31 Antibody (S04-4C1), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19699-100ul
This item is not returnable.
View return policy
Description
MRPS31 Monoclonal antibody specifically detects MRPS31 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MRPS31 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S04-4C1 | |
| Unconjugated | |
| 28S ribosomal protein S31, mitochondrial, IMOGN38Imogen 38, mitochondrial ribosomal protein S31, MRP-S31, S31mt | |
| A synthetic peptide of human MRPS31 (Uniprot # Q92665) | |
| 100 μg | |
| Endocrinology, Immunology | |
| 10240 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |