missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS33 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556469
This item is not returnable.
View return policy
Description
Immunogen sequence: MSSLSEYAFR MSRLSARLFG EVTRPTNSKS MKVVKLFSEL PLAKKKETYD WYPNHHTYAE LMQTLRFLGL YR Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. The 28S subunit of the mammalian mitoribosome may play a crucial and characteristic role in translation initiation. This gene encodes a 28S subunit protein that is one of the more highly conserved mitochondrial ribosomal proteins among mammals, Drosophila and C. elegans.
Specifications
| MRPS33 | |
| Polyclonal | |
| Unconjugated | |
| MRPS33 | |
| 28S ribosomal protein S33, mitochondrial; AI841153; CGI-139; Ganglioside-induced differentiation-associated-protein 3; Gdap3; mitochondrial ribosomal protein S33; Mitochondrial small ribosomal subunit protein mS33; Mrps33; MRP-S33; PTD003; rt33; S33mt | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51650 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y291 | |
| MRPS33 | |
| Recombinant protein corresponding to Human MRPS33. Recombinant protein control fragment (Product #RP-95584). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction