missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MRPS35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575457
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
Mammalian mitochondrial ribosomes (mitoribosomes) are responsible for protein synthesis within the mitochondrion. The mitoribosomes are composed of a 4:1 ratio of protein to RNA, with the proteins forming two subunits, the 28S subunit and the 39S subunit. Across species, the proteins that make up the mitoribosome subunits vary greatly in sequence, preventing easy recognition by sequence homology. MRP-S35 (mitochondrial ribosomal protein S35), also known as MDS023, MRPS28 or HDCMD11P, is a 323 amino acid protein that localizes to the mitochondrion, where it exists as a component of the 28S ribosomal subunit and works in conjunction with other MRPs to mediate protein synthesis. Existing as two alternatively spliced isoforms, MRP-S35 is encoded by a gene located on human chromosome 10, which houses over 1,200 genes and comprises nearly 4. 5% of the human genome.
Specifications
| MRPS35 | |
| Polyclonal | |
| Unconjugated | |
| MRPS35 | |
| 28S ribosomal protein S28, mitochondrial; 28S ribosomal protein S35, mitochondrial; HDCMD11P; MDS023; MDSO23; mitochondrial ribosomal protein S28; mitochondrial ribosomal protein S34; mitochondrial ribosomal protein S35; Mitochondrial small ribosomal subunit protein mS35; MRPS28; MRP-S28; Mrps35; MRP-S35; PSEC0213; S28mt; S35mt | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 60488 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P82673 | |
| MRPS35 | |
| Synthetic peptide corresponding to amino acids 220-270 of Human MRPS35. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction