missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS36 Polyclonal Antibody

MRPS36 Polyclonal Antibody
Brand: Invitrogen PA575468
This item is not returnable.
View return policy
Description
MRPS36 Polyclonal Antibody for IHC (P)
Specifications
| MRPS36 | |
| Polyclonal | |
| Unconjugated | |
| MRPS36 | |
| 1110018B13Rik; 28S ribosomal protein S36, mitochondrial; DC47; mitochondrial ribosomal protein S36; Mrps36; MRP-S36; RT36; S36mt | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 294696, 66128, 92259 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P82909, Q9CQX8 | |
| MRPS36 | |
| Synthetic peptide corresponding to the N-terminal of Human MRPS36. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction