missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 204.00 - € 481.00
Specifications
| Antigen | MRPS6 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662731
|
Novus Biologicals
NBP2-93563-0.02ml |
0.02 mL |
€ 204.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18627261
|
Novus Biologicals
NBP2-93563-0.1ml |
0.1 mL |
€ 481.00
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRPS6 Polyclonal antibody specifically detects MRPS6 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MRPS6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Endocrinology | |
| PBS (pH 7.3), 50% glycerol | |
| 64968 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| C21orf101, mitochondrial ribosomal protein S6, MRP-S6chromosome 21 open reading frame 101, RPMS628S ribosomal protein S6, mitochondrial, S6mt | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-125 of human MRPS6 (NP_115865.1). MPRYELALILKAMQRPETAATLKRTIEALMDRGAIVRDLENLGERALPYRISAHSQQHNRGGYFLVDFYAPTAAVESMVEHLSRDIDVIRGNIVKHPLTQELKECEGIVPVPLAEKLYSTKKRKK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title