missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPS7 Antibody (OTI5E11), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72789AF647
This item is not returnable.
View return policy
Description
MRPS7 Monoclonal specifically detects MRPS7 in Human samples. It is validated for Western Blot.
Specifications
| MRPS7 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MRPS7 | |
| Recombinant protein fragment corresponding to amino acids 38-242 of human MRPS7 (NP_057055) produced in E.coli. | |
| 0.1 mL | |
| 51081.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI5E11 | |
| Western Blot | |
| bMRP27a, bMRP-27a, mitochondrial ribosomal protein S7, MRP-S, MRP-S7,30S ribosomal protein S7 homolog, RPMS7,28S ribosomal protein S7, mitochondrial, RP-S7, S7mt | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction