missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRRF Antibody (OTI1D5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72790CL1
This item is not returnable.
View return policy
Description
MRRF Monoclonal antibody specifically detects MRRF in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| MRRF | |
| Monoclonal | |
| CoraFluor 1 | |
| mitochondrial, mitochondrial ribosome recycling factor, MTRRF | |
| Human recombinant protein fragment corresponding to amino acids 56-262 of human MRRF(NP_620132) produced in E.coli. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 92399 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot | |
| OTI1D5 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction