missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRRF Antibody (OTI1D5), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72790G
This item is not returnable.
View return policy
Description
MRRF Monoclonal specifically detects MRRF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MRRF | |
| Monoclonal | |
| DyLight 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| MRRF | |
| Recombinant protein fragment corresponding to amino acids 56-262 of human MRRF (NP_620132) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI1D5 | |
| Western Blot, Immunohistochemistry | |
| mitochondrial, mitochondrial ribosome recycling factor, MTRRF | |
| Mouse | |
| Affinity Purified | |
| DNA replication Transcription Translation and Splicing | |
| 92399.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction