missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRRF Monoclonal Antibody (OTI2A8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806309
This item is not returnable.
View return policy
Description
MRRF is responsible for the release of ribosomes from messenger RNA at the termination of protein biosynthesis, may increase the efficiency of translation by recycling ribosomes from one round of translation to another.Specifications
| MRRF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96E11 | |
| MRRF | |
| Human recombit protein fragment corresponding to amino acids 56-262 of human MRRF produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A8 | |
| Unconjugated | |
| MRRF | |
| MRFF, MTRRF, RRF | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 92399 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction