missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MRRF Polyclonal Antibody
GREENER_CHOICE

Product Code. 15759271 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15759271 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15759271 Supplier Invitrogen™ Supplier No. PA561458

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ALGLKCFRMV HPTFRNYLAA SIRPVSEVTL KTVHERQHGH RQYMAYSAVP VRHFATKKAK AKGKGQSQTR VNINAALVED IINLEEVNEE MKSV Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 71%.

Responsible for the release of ribosomes from messenger RNA at the termination of protein biosynthesis. May increase the efficiency of translation by recycling ribosomes from one round of translation to another. Ota T., Nat. Genet. 36:40-45(2004). Humphray S.J., Nature 429:369-374(2004). Zhang Y., Biochim. Biophys. Acta 1443:245-250(1998).
TRUSTED_SUSTAINABILITY

Specifications

Antigen MRRF
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mrrf
Gene Accession No. Q96E11
Gene Alias 2400002D02Rik; mitochondrial ribosome recycling factor; MRFF; MRRF; MTRRF; Ribosome-recycling factor, mitochondrial; ribosome-releasing factor, mitochondrial; RRF
Gene Symbols Mrrf
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MRRF. Recombinant protein control fragment (Product #RP-100158).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92399
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.