missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MRS 2365

Click to view available options
:
1mg
Description
MRS 2365 is a highly potent, selective P2Y1 receptor agonist (EC50 = 0.4 nM). Displays no activity at P2Y12 receptors and only very low agonist activity at P2Y13 receptors (at concentrations up to 1 μM). Increases the upregulation of NTPDase1 by ATPγS.
Specifications
Specifications
| Quantity | 1 mg |
| Product Type | MRS 2365 |
| Purity | >98% |
| Molecular Weight (g/mol) | 549.28 |
| Inhibitors | MRS 2365 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction