missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A1, anti-Human, Clone: rIGEL/773, Abnova™
Monoclonal Antibody
Brand: Abnova RAB00311.100ug
This item is not returnable.
View return policy
Specifications
| MS4A1 | |
| Monoclonal | |
| Unconjugated | |
| In 10 mM PBS (0.05% BSA and 0.05% azide). | |
| B1/Bp35/CD20/LEU-16/MGC3969/MS4A2/S7 | |
| Original antibody is raised against recombinant protein corresponding to full length human MS4A1. | |
| RUO | |
| 931 | |
| Store at 4°C. | |
| Liquid |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| rIGEL/773 | |
| Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| membrane-spanning 4-domains, subfamily A, member 1 | |
| MS4A1 | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction