missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MS4A14 Antibody (SR4233), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 316.00 - € 382.00
Specifications
| Antigen | MS4A14 |
|---|---|
| Clone | SR4233 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041566
|
Novus Biologicals™
NBP3-27828-100ul |
100 μL |
€ 382.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041606
|
Novus Biologicals™
NBP3-27828-50ul |
50 μL |
€ 316.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MS4A14 Monoclonal antibody specifically detects MS4A14 in Human samples. It is validated for Western BlotSpecifications
| MS4A14 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 84689 | |
| IgG | |
| Affinity purified |
| SR4233 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DKFZp434H092, FLJ32856, membrane-spanning 4-domains subfamily A member 14, membrane-spanning 4-domains, subfamily A, member 14, membrane-spanning 4-domains, subfamily A, member 16, MGC104289, MGC49828, MS4A13, MS4A13 protein, MS4A16, NYD-SP21, testes development-related NYD-SP21, Testis development protein NYD-SP21 | |
| A synthesized peptide derived from human MS4A14 (Uniprot #: Q96JA4) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title