Learn More
Invitrogen™ MS4A14 Recombinant Rabbit Monoclonal Antibody (24GB3545)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557083
 View more versions of this product
Product Code. 30276255
  • € 514.00 / 100µL
Description
MS4A14 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
May be involved in signal transduction as a component of a multimeric receptor complex.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MS4A14
Recombinant Monoclonal
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q96JA4
MS4A14
A synthesized peptide derived from human MS4A14 (aa 150-280).
100 μL
Primary
Human, Mouse
Antibody
IgG
Western Blot, Immunocytochemistry
24GB3545
Unconjugated
MS4A14
membrane spanning 4-domains A14; membrane-spanning 4-domains subfamily A member 14; membrane-spanning 4-domains, subfamily A, member 14; membrane-spanning 4-domains, subfamily A, member 16; MS4A13; MS4A13 protein; MS4A14; MS4A16; NYD-SP21; testes development-related NYD-SP21; testis development protein NYD-SP21
Rabbit
Affinity chromatography
RUO
383435, 84689
-20°C