missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A15, Mouse anti-Human, Clone: 1D4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00219995-M03.100ug
This item is not returnable.
View return policy
Description
Sequence: MVRRGHVGIFFIEGGVPFWGGACFIISGSLSVAAEKNHTSCLVRSSLGTNILSVMAAFAGTAILLMDFGVTNRDVDRGYLAVLTIFTVLEFFTAVIAMHFGCQAIHAQASAPVIFLPNAFSADFNIPSPAASAPPAYDNVAYAQGVVSpecifications
| MS4A15 | |
| Monoclonal | |
| Unconjugated | |
| membrane-spanning 4-domains, subfamily A, member 15 | |
| FLJ34527/MGC35295 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1D4 | |
| In 1x PBS, pH 7.4 | |
| BC031610 | |
| MS4A15 | |
| MGC35295 (AAH31610.1, 1 a.a. ∼ 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 219995 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction