missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MS4A4A Recombinant Mouse Monoclonal Antibody (5C12)

Mouse Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA548222
This item is not returnable.
View return policy
Description
Specificity: This antibody is specific for human extracellular MS4A4A.
This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. Alternative splicing of this gene results in several transcript variants; however, not all transcripts have been fully described.
Specifications
| MS4A4A | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% ProClin 300 | |
| Q96JQ5 | |
| MS4A4A | |
| This antibody was generated by immunizing C57BL/6 mice against mouse cell line 300-19 expressing transfected human membrane-spanning 4-domains subfamily A member 4A (MS4A4A)-GFP. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 λ |
| Flow Cytometry, Immunoprecipitation | |
| 5C12 | |
| Unconjugated | |
| MS4A4A | |
| 4SPAN1; CD20 antigen-like 1; CD20L1; CD20-L1; Fc epsilon receptor beta subunit homolog; four-span transmembrane protein 1; HDCME31P; membrane spanning 4-domains A4A; membrane-spanning 4-domains subfamily A member 4A; membrane-spanning 4-domains, subfamily A, member 4; membrane-spanning 4-domains, subfamily A, member 4A; MS4A4; MS4A4A; MS4A7 | |
| Mouse | |
| Protein A | |
| RUO | |
| 51338 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction