missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MS4A5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560486
This item is not returnable.
View return policy
Description
Immunogen sequence: VFLVFPPEIT ASEYESTELS ATTF Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.
May be involved in signal transduction as a component of a multimeric receptor complex.
Specifications
| MS4A5 | |
| Polyclonal | |
| Unconjugated | |
| MS4A5 | |
| CD20 antigen-like 2; CD20L2; CD20-L2; membrane spanning 4-domains A5; membrane-spanning 4-domains subfamily A member 5; membrane-spanning 4-domains, subfamily A, member 5; MS4A5; Testis-expressed transmembrane protein 4; testis-expressed transmembrane-4 protein; TETM4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 64232 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H3V2 | |
| MS4A5 | |
| Recombinant protein corresponding to Human MS4A5. Recombinant protein control fragment (Product #RP-100439). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction