missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSC Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549769
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total MSC protein.
MyoD-E or musculin-E heterodimers bind avidly to consensus E box motifs, which are functionally important elements in the promoter regions of many musclespecific terminal differentiation genes. MyoD complexes potently induce transcriptional activation, while musculin complexes bind adjacent to MyoD DNA-binding regions to represses MyoD activity, which then results in the delayed expression of muscle-specific genes. Musculin is highly expressed in undifferentiated and proliferating myoblasts in culture, and its expression is down regulated during myogenesis and at the onset of terminal differentiation.
Specifications
| MSC | |
| Polyclonal | |
| Unconjugated | |
| MSC | |
| ABF1; ABF-1; Activated B-cell factor 1; activated B-cell factor 1, homolog of mouse musculin; activated B-cell factor-1; bHLHa22; Class A basic helix-loop-helix protein 22; Msc; Musculin; myogenic repressor; Myor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17681, 9242 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O60682, O88940 | |
| MSC | |
| Synthesized peptide derived from internal of human Musculin. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction