missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH2 Antibody (rMSH2/6548), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08956APC
This item is not returnable.
View return policy
Description
MSH2 Monoclonal antibody specifically detects MSH2 in Human, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
Specifications
| MSH2 | |
| Monoclonal | |
| APC | |
| COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2 | |
| Recombinant full-length human MSH2 protein (Uniprot: IDP43246) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors | |
| 4436 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry (Paraffin) | |
| rMSH2/6548 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Canine, Feline | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction