missing translation for 'onlineSavingsMsg'
Learn More

MSH2 Antibody (rMSH2/6548), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-08956DL594

Product Code. 18548116

  • ONLINE EXCLUSIVE PRICE
    € 643.00
    € 607.95 / 100µL
    Save € 35.05
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MSH2 Monoclonal antibody specifically detects MSH2 in Human, Canine, Feline samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MSH2
Monoclonal
DyLight 594
COCA1, DNA mismatch repair protein Msh2, FCC1, hMSH2, HNPCC, HNPCC1mutS (E. coli) homolog 2 (colon cancer, nonpolyposis type 1), LCFS2, mutS homolog 2, colon cancer, nonpolyposis type 1 (E. coli), MutS protein homolog 2
Recombinant full-length human MSH2 protein (Uniprot: IDP43246)
100 μg
Breast Cancer, Cancer, Core ESC Like Genes, DNA Repair, Mismatch Repair, Stem Cell Markers, Tumor Suppressors
4436
Store at 4°C in the dark.
IgG2a κ
Western Blot, Immunohistochemistry (Paraffin)
rMSH2/6548
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human, Canine, Feline
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.