Learn More
MSH3 Rabbit, anti-Human, Clone: 1J23, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand:  Proteintech 82845-3-RR-20UL
 View more versions of this product
Product Code. 18035255
  • € 149.00 / 20µL
Description
The MSH3 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human MSH3. This antibody recognizes human antigen. The MSH3 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
MSH3
1J23
Divergent upstream protein, DUC1, DUG, DUP, hMSH3, Mismatch repair protein 1, MRP1, MSH3, mutS homolog 3 (E. coli)
20 μL
4437
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, Immunofluorescence, ELISA
1000 μg/mL
Rabbit
Primary
Human
150 kDa