missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MSH4 Polyclonal antibody specifically detects MSH4 in Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MSH4 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500 - 1:2000 |
| Formulation | PBS (pH 7.3), 50% glycerol |
| Gene Alias | hMSH4, mutS (E. coli) homolog 4, mutS homolog 4 (E. coli), mutS protein homolog 4 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 697-936 of human MSH4 (NP_002431.2). QIMAQIGSYVPAEYSSFRIAKQIFTRISTDDDIETNSSTFMKEMKEIAYILHNANDKSLILIDELGRGTNTEEGIGICYAVCEYLLSLKAFTLFATHFLELCHIDALYPNVENMHFEVQHVKNTSRNKEAILYTYKLSKGLTEEKNYGLKAAEVSSLPPSIVLDAKEITTQITRQILQNQRSTPEMERQRAVYHLATRLVQTARNSQLDPDSLRIYLSNLKKKYKEDFPRTEQVPEKTEE |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?