missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH6 Antibody (MSH6/3086), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79846MFV450
This item is not returnable.
View return policy
Description
MSH6 Monoclonal antibody specifically detects MSH6 in Human samples. It is validated for Flow Cytometry
Specifications
| MSH6 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| MSH6/3086 | |
| Flow Cytometry | |
| DNA mismatch repair protein Msh6, G/T mismatch-binding protein, GTBPHNPCC5, GTMBP, hMSH6, HSAP, mutS (E. coli) homolog 6, mutS homolog 6 (E. coli), MutS-alpha 160 kDa subunit, p160, sperm-associated protein | |
| Recombinant fragment of human MSH6 protein (around aa 374-540) (exact sequence is proprietary) (Uniprot: P52701) | |
| 0.1 mL | |
| Breast Cancer, Cancer, DNA Repair, Mismatch Repair | |
| 2956 | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction