missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH6 Antibody (MSH6/8338R), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24204MFV610
This item is not returnable.
View return policy
Description
MSH6 Monoclonal antibody specifically detects MSH6 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| MSH6 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| DNA mismatch repair protein Msh6, G/T mismatch-binding protein, GTBPHNPCC5, GTMBP, hMSH6, HSAP, mutS (E. coli) homolog 6, mutS homolog 6 (E. coli), MutS-alpha 160 kDa subunit, p160, sperm-associated protein | |
| Recombinant fragment (around aa 1-200) of human MSH6 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Breast Cancer, Cancer, DNA Repair, Mismatch Repair | |
| 2956 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MSH6/8338R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction