missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18120-1-AP-20UL
This item is not returnable.
View return policy
Description
The DNA mismatch repair MSH2/MSH6 heterodimer is responsible for the detection of DNA damage, and may promote apoptosis in normal cells, thus protecting mammals from ultraviolet-induced maligt transformation and hence the prevention of skin cancer.Specifications
| MSH6 | |
| Polyclonal | |
| Unconjugated | |
| MSH6 | |
| G/T mismatch binding protein, GTBP, GTMBP, hMSH6, HNPCC5, HSAP, MSH6, MutS alpha 160 kDa subunit, mutS homolog 6 (E. coli), p160 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100360342, 17688, 2956 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry, Western Blot | |
| 0.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52701, P54276 | |
| Msh6 | |
| MSH6 Fusion Protein Ag12645 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction