missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSL2 Monoclonal Antibody (OTI3B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811188
This item is not returnable.
View return policy
Description
Component of histone acetyltransferase complex responsible for the majority of histone H4 acetylation at lysine 16 which is implicated in the formation of higher-order chromatin structure. Acts as an E3 ubiquitin ligase that promotes monoubiquitination of histone H2B at ′Lys-35′ (H2BK34Ub), but not that of H2A. This activity is greatly enhanced by heterodimerization with MSL1. H2B ubiquitination in turn stimulates histine H3 methylation at ′Lys-4′ (H3K4me) and ′Lys-79′ (H3K79me) and leads to gene activation, including that of HOXA9 and MEIS1.Specifications
| MSL2 | |
| Monoclonal | |
| Unconjugated | |
| MSL2 | |
| MSL-2, MSL2L1, RNF184 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 55167 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI3B11 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9HCI7 | |
| MSL2 | |
| Full length human recombit protein of human MSL2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction